Assay Design



Gene Transcripts

Gene Symbol
DFFB
Entrez Gene ID
512730
Gene Name
DNA fragmentation factor subunit beta
Gene Aliases
-
Chromosome Location
Chr.16: 49213722 - 49228162 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035109.1 NP_001030281.1 1 - 2 235 107
GenBank mRNA BC149123.1 - 1 - 2 172 107
BT021807.1 - 1 - 2 235 107

View Full Product Details