Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035113.1 | NP_001030285.1 | 1 - 2 | 102 | 65 |
GenBank mRNA | BC105255.1 | - | 1 - 2 | 101 | 65 |
BT021527.1 | - | 1 - 2 | 102 | 65 | |
BT021540.1 | - | 1 - 2 | 82 | 65 | |
BT021619.1 | - | 1 - 2 | 33 | 65 | |
BT021755.1 | - | 1 - 2 | 81 | 65 |