Assay Design



Gene Transcripts

Gene Symbol
NKG7
Entrez Gene ID
513006
Gene Name
natural killer cell granule protein 7
Gene Aliases
-
Chromosome Location
Chr.18: 57439878 - 57442652 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035279.1 NP_001030356.1 3 - 4 600 61
GenBank mRNA BC105575.1 - 3 - 4 629 61
BT021488.1 - 3 - 4 600 61

View Full Product Details