Assay Design



Gene Transcripts

Gene Symbol
B9D2
Entrez Gene ID
616609
Gene Name
B9 domain containing 2
Gene Aliases
-
Chromosome Location
Chr.18: 50509250 - 50516883 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040593.2 NP_001035683.1 2 - 3 225 108
GenBank mRNA AY911338.1 - 2 - 3 226 108

View Full Product Details