Assay Design



Gene Transcripts

Gene Symbol
PRXL2B
Entrez Gene ID
617001
Gene Name
peroxiredoxin like 2B
Gene Aliases
C16H1orf93, C1orf93, FAM213B
Chromosome Location
Chr.16: 50307838 - 50310952 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040598.1 NP_001035688.1 6 - 7 673 59
GenBank mRNA BC114900.1 - 6 - 7 592 59
BT021811.1 - 6 - 7 673 59

View Full Product Details