Assay Design



Gene Transcripts

Gene Symbol
GADD45B
Entrez Gene ID
618405
Gene Name
growth arrest and DNA damage inducible beta
Gene Aliases
-
Chromosome Location
Chr.7: 21118048 - 21120156 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001040604.1 NP_001035694.1 2 - 3 356 71
GenBank mRNA BC113347.1 - 2 - 3 370 71
BT021015.1 - 2 - 3 356 71

View Full Product Details