Assay Design



Gene Transcripts

Gene Symbol
BRCC3
Entrez Gene ID
519513
Gene Name
BRCA1/BRCA2-containing complex subunit 3
Gene Aliases
CXorf53
Chromosome Location
Chr.X: 35885699 - 35964218 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075790.1 NP_001069258.1 6 - 7 511 123
GenBank mRNA BC142193.1 - 6 - 7 511 123
BT021737.1 - 6 - 7 511 123

View Full Product Details