Assay Design



Gene Transcripts

Gene Symbol
BRCC3
Entrez Gene ID
519513
Gene Name
BRCA1/BRCA2-containing complex subunit 3
Gene Aliases
CXorf53
Chromosome Location
Chr.X: 35885699 - 35964218 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001075790.1 NP_001069258.1 9 - 10 737 80
GenBank mRNA BC142193.1 - 10 - 11 812 80
BT021737.1 - 9 - 10 737 80

View Full Product Details