Assay Design



Gene Transcripts

Gene Symbol
TEX30
Entrez Gene ID
613895
Gene Name
testis expressed 30
Gene Aliases
C12H13orf27, C13orf27
Chromosome Location
Chr.12: 79008124 - 79014993 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001076295.2 NP_001069763.2 1 - 2 116 60
GenBank mRNA BC102911.1 - 1 - 2 117 60
EH156727.1 - 1 - 2 112 60

View Full Product Details