Assay Design



Gene Transcripts

Gene Symbol
CLYBL
Entrez Gene ID
533186
Gene Name
citramalyl-CoA lyase
Gene Aliases
-
Chromosome Location
Chr.12: 76430699 - 76628229 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080296.1 NP_001073765.1 2 - 3 284 77
GenBank mRNA BC147857.1 - 2 - 3 263 77
BT021750.1 - 2 - 3 284 77

View Full Product Details