Assay Design



Gene Transcripts

Gene Symbol
CLYBL
Entrez Gene ID
533186
Gene Name
citramalyl-CoA lyase
Gene Aliases
-
Chromosome Location
Chr.12: 76430699 - 76628229 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080296.1 NP_001073765.1 4 - 5 561 61
GenBank mRNA BT021750.1 - 4 - 5 561 61

View Full Product Details