Assay Design



Gene Transcripts

Gene Symbol
MRPL3
Entrez Gene ID
614906
Gene Name
mitochondrial ribosomal protein L3
Gene Aliases
-
Chromosome Location
Chr.1: 138384515 - 138441535 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080317.2 NP_001073786.1 9 - 10 1012 118
GenBank mRNA BC103047.1 - 9 - 10 1013 118

View Full Product Details