Assay Design



Gene Transcripts

Gene Symbol
APOH
Entrez Gene ID
281006
Gene Name
apolipoprotein H
Gene Aliases
-
Chromosome Location
Chr.19: 62611657 - 62624442 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173992.2 NP_776417.1 3 - 4 371 95
GenBank mRNA BC118364.1 - 3 - 4 95
L07303.2 - 3 - 4 374 95
X60065.1 - - 95

View Full Product Details