Assay Design



Gene Transcripts

Gene Symbol
IL13
Entrez Gene ID
281247
Gene Name
interleukin 13
Gene Aliases
-
Chromosome Location
Chr.7: 21721568 - 21723568 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174089.1 NP_776514.1 2 - 3 254 76
GenBank mRNA AF072807.1 - 2 - 3 184 76
EU276077.1 - 2 - 3 227 76

View Full Product Details