Assay Design
Gene Transcripts
- Gene Symbol
- CHRNB4
- Entrez Gene ID
-
282181
- Gene Name
- cholinergic receptor nicotinic beta 4 subunit
- Gene Aliases
-
-
- Chromosome Location
-
Chr.21: 30996280 - 31015137 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174517.1 |
NP_776942.1 |
4 - 5 |
365 |
68 |
GenBank mRNA |
AF487465.1 |
- |
4 - 5 |
365 |
68 |
View Full Product Details