Assay Design
Gene Transcripts
- Gene Symbol
- PTHLH
- Entrez Gene ID
-
286767
- Gene Name
- parathyroid hormone like hormone
- Gene Aliases
-
PTHrP
- Chromosome Location
-
Chr.5: 81841203 - 81858767 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_174753.1 |
NP_777178.1 |
1 - 2 |
115 |
88 |
GenBank mRNA |
AB097837.1 |
- |
1 - 2 |
115 |
88 |
View Full Product Details