Assay Design



Gene Transcripts

Gene Symbol
PTHLH
Entrez Gene ID
286767
Gene Name
parathyroid hormone like hormone
Gene Aliases
PTHrP
Chromosome Location
Chr.5: 81841203 - 81858767 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_174753.1 NP_777178.1 2 - 3 238 117
GenBank mRNA AB097837.1 - 2 - 3 238 117
BC149411.1 - 3 - 4 226 117

View Full Product Details