Assay Design



Gene Transcripts

Gene Symbol
CTBP2
Entrez Gene ID
281730
Gene Name
C-terminal binding protein 2
Gene Aliases
-
Chromosome Location
Chr.26: 44545693 - 44714474 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_175712.1 NP_783643.1 8 - 9 2994 67
GenBank mRNA AF222713.2 - 8 - 9 2994 67
BC120058.1 - 10 - 11 1445 67

View Full Product Details