Assay Design
Gene Transcripts
- Gene Symbol
- IL21
- Entrez Gene ID
-
378475
- Gene Name
- interleukin 21
- Gene Aliases
-
-
- Chromosome Location
-
Chr.17: 35063113 - 35070244 on Build ARS-UCD1.2
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_198832.1 |
NP_942129.1 |
2 - 3 |
199 |
122 |
GenBank mRNA |
AB073021.1 |
- |
2 - 3 |
199 |
122 |
View Full Product Details