Assay Design



Gene Transcripts

Gene Symbol
CENPH
Entrez Gene ID
505284
Gene Name
centromere protein H
Gene Aliases
-
Chromosome Location
Chr.20: 10483632 - 10502446 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034236.2 NP_001029408.1 6 - 7 517 143
GenBank mRNA BC102350.1 - 6 - 7 518 143

View Full Product Details