Assay Design



Gene Transcripts

Gene Symbol
PNRC1
Entrez Gene ID
507736
Gene Name
proline rich nuclear receptor coactivator 1
Gene Aliases
-
Chromosome Location
Chr.9: 61030512 - 61034542 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034308.2 NP_001029480.1 1 - 2 683 106
GenBank mRNA BC105159.1 - 1 - 2 684 106

View Full Product Details