Assay Design



Gene Transcripts

Gene Symbol
POLE3
Entrez Gene ID
510678
Gene Name
DNA polymerase epsilon 3, accessory subunit
Gene Aliases
-
Chromosome Location
Chr.8: 102655928 - 102658552 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034390.1 NP_001029562.1 3 - 4 333 84
GenBank mRNA BC102772.1 - 3 - 4 333 84
BT030577.1 - 3 - 4 307 84

View Full Product Details