Assay Design



Gene Transcripts

Gene Symbol
DYNLRB1
Entrez Gene ID
513342
Gene Name
dynein light chain roadblock-type 1
Gene Aliases
-
Chromosome Location
Chr.13: 63888186 - 63905673 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034442.2 NP_001029614.1 3 - 4 290 105
GenBank mRNA BC102138.1 - 3 - 4 292 105

View Full Product Details