Assay Design



Gene Transcripts

Gene Symbol
FBP1
Entrez Gene ID
513483
Gene Name
fructose-bisphosphatase 1
Gene Aliases
-
Chromosome Location
Chr.8: 81068093 - 81098919 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034447.2 NP_001029619.1 6 - 7 929 122
GenBank mRNA BC102974.1 - 6 - 7 931 122
DQ520945.1 - 6 - 7 888 122

View Full Product Details