Assay Design



Gene Transcripts

Gene Symbol
MLF2
Entrez Gene ID
513843
Gene Name
myeloid leukemia factor 2
Gene Aliases
-
Chromosome Location
Chr.5: 103691259 - 103696646 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034456.1 NP_001029628.1 8 - 9 946 117
GenBank mRNA BC104523.1 - 8 - 9 946 117

View Full Product Details