Assay Design



Gene Transcripts

Gene Symbol
MED4
Entrez Gene ID
515299
Gene Name
mediator complex subunit 4
Gene Aliases
-
Chromosome Location
Chr.12: 17896752 - 17938657 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034486.1 NP_001029658.1 2 - 3 192 92
GenBank mRNA BC103312.1 - 2 - 3 192 92

View Full Product Details