Assay Design



Gene Transcripts

Gene Symbol
SPSB3
Entrez Gene ID
515661
Gene Name
splA/ryanodine receptor domain and SOCS box containing 3
Gene Aliases
-
Chromosome Location
Chr.25: 1355611 - 1361300 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034506.1 NP_001029678.1 2 - 3 167 120
GenBank mRNA BC104512.1 - 2 - 3 167 120

View Full Product Details