Assay Design



Gene Transcripts

Gene Symbol
DDX19A
Entrez Gene ID
529929
Gene Name
DEAD-box helicase 19A
Gene Aliases
-
Chromosome Location
Chr.18: 1734904 - 1753496 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034571.2 NP_001029743.1 5 - 6 499 64
GenBank mRNA BC103093.1 - 5 - 6 500 64
BT021652.1 - 5 - 6 471 64

View Full Product Details