Assay Design



Gene Transcripts

Gene Symbol
PDZK1
Entrez Gene ID
534439
Gene Name
PDZ domain containing 1
Gene Aliases
-
Chromosome Location
Chr.3: 21648117 - 21695753 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034611.2 NP_001029783.1 - 73 84
GenBank mRNA BC102213.1 - - 74 84

View Full Product Details