Assay Design



Gene Transcripts

Gene Symbol
HPX
Entrez Gene ID
534509
Gene Name
hemopexin
Gene Aliases
-
Chromosome Location
Chr.15: 46606398 - 46615970 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034612.2 NP_001029784.1 4 - 5 363 106
GenBank mRNA BC102687.1 - 4 - 5 364 106

View Full Product Details