Assay Design



Gene Transcripts

Gene Symbol
STMN2
Entrez Gene ID
534991
Gene Name
stathmin 2
Gene Aliases
-
Chromosome Location
Chr.14: 42811857 - 42870107 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034622.3 NP_001029794.1 4 - 5 853 87
GenBank mRNA BC105211.1 - 4 - 5 589 87

View Full Product Details