Assay Design



Gene Transcripts

Gene Symbol
SLC35A1
Entrez Gene ID
536838
Gene Name
solute carrier family 35 member A1
Gene Aliases
-
Chromosome Location
Chr.9: 62413958 - 62445501 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034637.2 NP_001029809.1 4 - 5 534 89
GenBank mRNA BC102765.1 - 4 - 5 535 89

View Full Product Details