Assay Design



Gene Transcripts

Gene Symbol
PSMA2
Entrez Gene ID
539141
Gene Name
proteasome 20S subunit alpha 2
Gene Aliases
-
Chromosome Location
Chr.4: 78127061 - 78137710 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034662.2 NP_001029834.1 6 - 7 559 127
GenBank mRNA BC102206.1 - 6 - 7 560 127

View Full Product Details