Assay Design



Gene Transcripts

Gene Symbol
GDE1
Entrez Gene ID
539978
Gene Name
glycerophosphodiester phosphodiesterase 1
Gene Aliases
MIR16
Chromosome Location
Chr.25: 17101509 - 17126996 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034686.2 NP_001029858.1 4 - 5 711 87
GenBank mRNA BC102273.1 - 4 - 5 712 87

View Full Product Details