Assay Design



Gene Transcripts

Gene Symbol
OTUB1
Entrez Gene ID
540984
Gene Name
OTU deubiquitinase, ubiquitin aldehyde binding 1
Gene Aliases
-
Chromosome Location
Chr.29: 42315651 - 42323036 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034719.2 NP_001029891.1 4 - 5 357 72
GenBank mRNA BC102210.1 - 4 - 5 359 72

View Full Product Details