Assay Design



Gene Transcripts

Gene Symbol
AVPI1
Entrez Gene ID
614488
Gene Name
arginine vasopressin induced 1
Gene Aliases
-
Chromosome Location
Chr.26: 18846870 - 18855141 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034760.2 NP_001029932.1 1 - 2 358 70
GenBank mRNA BC102744.1 - 1 - 2 359 70

View Full Product Details