Assay Design



Gene Transcripts

Gene Symbol
SSMEM1
Entrez Gene ID
615577
Gene Name
serine rich single-pass membrane protein 1
Gene Aliases
C4H7orf45
Chromosome Location
Chr.4: 93985493 - 93995178 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034785.2 NP_001029957.1 1 - 2 230 109
GenBank mRNA BC102591.1 - 1 - 2 231 109

View Full Product Details