Assay Design



Gene Transcripts

Gene Symbol
STMN1
Entrez Gene ID
616317
Gene Name
stathmin 1
Gene Aliases
-
Chromosome Location
Chr.2: 127152984 - 127183810 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034790.2 NP_001029962.1 3 - 4 277 125
GenBank mRNA BC102453.1 - 3 - 4 278 125

View Full Product Details