Assay Design



Gene Transcripts

Gene Symbol
GALM
Entrez Gene ID
616676
Gene Name
galactose mutarotase
Gene Aliases
-
Chromosome Location
Chr.11: 21021100 - 21075769 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001034795.1 NP_001029967.1 1 - 2 236 70
GenBank mRNA BC102446.1 - 1 - 2 236 70
BT020690.1 - 1 - 2 253 70

View Full Product Details