Assay Design



Gene Transcripts

Gene Symbol
OMA1
Entrez Gene ID
506223
Gene Name
OMA1 zinc metallopeptidase
Gene Aliases
-
Chromosome Location
Chr.3: 87479235 - 87572929 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035033.2 NP_001030205.1 4 - 5 974 113
GenBank mRNA BC102774.1 - 4 - 5 975 113

View Full Product Details