Assay Design



Gene Transcripts

Gene Symbol
OMA1
Entrez Gene ID
506223
Gene Name
OMA1 zinc metallopeptidase
Gene Aliases
-
Chromosome Location
Chr.3: 87479235 - 87572929 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035033.2 NP_001030205.1 7 - 8 1286 73
GenBank mRNA BC102774.1 - 7 - 8 1287 73

View Full Product Details