Assay Design



Gene Transcripts

Gene Symbol
TMA16
Entrez Gene ID
506277
Gene Name
translation machinery associated 16 homolog
Gene Aliases
C6H4orf43
Chromosome Location
Chr.6: 2111634 - 2141474 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035034.2 NP_001030206.1 6 - 7 465 116
GenBank mRNA BC102541.1 - 6 - 7 466 116

View Full Product Details