Assay Design



Gene Transcripts

Gene Symbol
ANP32B
Entrez Gene ID
509685
Gene Name
acidic nuclear phosphoprotein 32 family member B
Gene Aliases
-
Chromosome Location
Chr.8: 63155268 - 63179623 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035074.1 NP_001030246.1 2 - 3 436 142
GenBank mRNA BC102954.1 - 2 - 3 436 142

View Full Product Details