Assay Design



Gene Transcripts

Gene Symbol
ITM2B
Entrez Gene ID
510575
Gene Name
integral membrane protein 2B
Gene Aliases
Bri, imBRI2
Chromosome Location
Chr.12: 18036781 - 18061776 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035093.2 NP_001030265.1 2 - 3 404 133
GenBank mRNA BC102307.1 - 2 - 3 405 133

View Full Product Details