Assay Design



Gene Transcripts

Gene Symbol
TSR2
Entrez Gene ID
511210
Gene Name
TSR2 ribosome maturation factor
Gene Aliases
-
Chromosome Location
Chr.X: 91960654 - 91965628 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035097.2 NP_001030269.1 2 - 3 187 63
GenBank mRNA BC102505.1 - 2 - 3 188 63

View Full Product Details