Assay Design



Gene Transcripts

Gene Symbol
EDF1
Entrez Gene ID
515380
Gene Name
endothelial differentiation related factor 1
Gene Aliases
-
Chromosome Location
Chr.11: 105948291 - 105952197 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035307.2 NP_001030384.1 4 - 5 485 66
GenBank mRNA BC102246.1 - 4 - 5 487 66

View Full Product Details