Assay Design



Gene Transcripts

Gene Symbol
MRPS10
Entrez Gene ID
515885
Gene Name
mitochondrial ribosomal protein S10
Gene Aliases
-
Chromosome Location
Chr.23: 15918487 - 15932587 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035314.2 NP_001030391.1 6 - 7 533 110
GenBank mRNA BC102893.1 - 6 - 7 534 110

View Full Product Details