Assay Design



Gene Transcripts

Gene Symbol
LIPC
Entrez Gene ID
535192
Gene Name
lipase C, hepatic type
Gene Aliases
-
Chromosome Location
Chr.10: 51696718 - 51883306 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035410.2 NP_001030487.1 6 - 7 1081 92
GenBank mRNA BC103072.1 - 6 - 7 1081 92

View Full Product Details