Assay Design



Gene Transcripts

Gene Symbol
MRPL22
Entrez Gene ID
614639
Gene Name
mitochondrial ribosomal protein L22
Gene Aliases
-
Chromosome Location
Chr.7: 66108715 - 66147126 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035454.2 NP_001030531.1 2 - 3 100 75
GenBank mRNA BC102665.1 - 2 - 3 101 75

View Full Product Details