Assay Design



Gene Transcripts

Gene Symbol
CHCHD3
Entrez Gene ID
616477
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 3
Gene Aliases
-
Chromosome Location
Chr.4: 96468819 - 96761374 on Build ARS-UCD1.2
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001035475.1 NP_001030552.1 7 - 8 811 106
GenBank mRNA BC102347.1 - 7 - 8 811 106
BT020987.1 - 7 - 8 778 106

View Full Product Details